missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OX40/TNFRSF4 Antibody (2553D) [DyLight 680], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB10563Y
This item is not returnable.
View return policy
Description
OX40/TNFRSF4 Monoclonal antibody specifically detects OX40/TNFRSF4 in Human samples. It is validated for Flow Cytometry, Neutralization, CyTOF
Specifications
| OX40/TNFRSF4 | |
| Monoclonal | |
| DyLight 680 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Neutralization, CyTOF | |
| 2553D | |
| Flow Cytometry, Neutralization, CyTOF-ready | |
| ACT35 antigen, ACT35ATC35 antigen, CD134, CD134 antigen, OX40 cell surface antigen, OX40 homologue, OX40L receptor, OX40lymphoid activation antigene ACT35, TAX transcriptionally-activated glycoprotein 1 receptor, tax-transcriptionally activated glycoprotein 1 receptor, tumor necrosis factor receptor superfamily member 4, tumor necrosis factor receptor superfamily, member 4, TXGP1LOX40 antigen | |
| Mouse myeloma cell line NS0-derived human OX40/TNFRSF4, Leu29-Ala216, Accession # P34389 | |
| 0.1 mL | |
| Adaptive Immunity, B Cell Development and Differentiation Markers, Cancer, Cell Cycle and Replication, Immunology, Myeloid derived Suppressor Cell, Signal Transduction | |
| 7293 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction