missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ OX40/TNFRSF4 Antibody (BMS-986178) - Humanized, Novus Biologicals™
Shop All Bio Techne ProductsDescription
OX40/TNFRSF4 Monoclonal antibody specifically detects OX40/TNFRSF4 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
Specifications
| Antigen | OX40/TNFRSF4 |
| Applications | Flow Cytometry, ELISA, Functional Assay |
| Classification | Monoclonal |
| Clone | BMS-986178 |
| Conjugate | Unconjugated |
| Formulation | Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) |
| Gene Accession No. | P43489 |
| Gene Alias | ACT35 antigen, ACT35ATC35 antigen, CD134, CD134 antigen, OX40 cell surface antigen, OX40 homologue, OX40L receptor, OX40lymphoid activation antigene ACT35, TAX transcriptionally-activated glycoprotein 1 receptor, tax-transcriptionally activated glycoprotein 1 receptor, tumor necrosis factor receptor superfamily member 4, tumor necrosis factor receptor superfamily, member 4, TXGP1LOX40 antigen |
| Host Species | Human |
| Immunogen | TNFRSF4/ OX40/ CD134 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?