missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OX40/TNFRSF4 Antibody (OX-86), Biotin, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NB100-64847B
This item is not returnable.
View return policy
Description
OX40/TNFRSF4 Monoclonal specifically detects OX40/TNFRSF4 in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
| OX40/TNFRSF4 | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| TNFRSF4 | |
| CHO cells transfected with murine OX40 | |
| 0.125 mL | |
| Cancer, Cell Cycle and Replication, Immunology | |
| 7293 | |
| Mouse | |
| Purified |
| Immunohistochemistry (Frozen) | |
| OX-86 | |
| Immunohistochemistry-Frozen | |
| ACT35 antigen, ACT35ATC35 antigen, CD134, CD134 antigen, OX40 cell surface antigen, OX40 homologue, OX40L receptor, OX40lymphoid activation antigene ACT35, TAX transcriptionally-activated glycoprotein 1 receptor, tax-transcriptionally activated glycoprotein 1 receptor, tumor necrosis factor receptor superfamily member 4, tumor necrosis factor receptor superfamily, member 4, TXGP1LOX40 antigen | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| NB100-64847 recognises murine OX40 (CD134), a cell surface antigen expressed only by activated T lymphocytes. | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction