Learn More
Tocris Bioscience™ OXA 06 dihydrochloride
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
OXA 06 dihydrochloride is a potent ROCK inhibitor (IC50 = 10 nM). Suppresses pMYPT1 and pCofilin levels in non-small cell lung carcinoma (NSCLC) cell lines. Also inhibits anchorage-independent growth of NSCLC cell lines in vitro.
Specifications
Specifications
| CAS | 1825455-91-1 |
| Quantity | 10 mg |
| Product Type | OXA 06 dihydrochloride |
| Purity | >98% |
| Molecular Weight (g/mol) | 404.31 |
| Inhibitors | OXA 06 dihydrochloride |