missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OXA1L Polyclonal Antibody

OXA1L Polyclonal Antibody
Brand: Invitrogen PA542895
This item is not returnable.
View return policy
Description
OXA1L Polyclonal Antibody for Western Blot
Specifications
| OXA1L | |
| Polyclonal | |
| Unconjugated | |
| OXA1L | |
| 1810020M02Rik; AI894287; Hsa; hypothetical protein LOC520335; LOW QUALITY PROTEIN: mitochondrial inner membrane protein OXA1L; mitochondrial inner membrane protein OXA1L; OXA1; OXA1Hs; Oxa1l; OXA1L mitochondrial inner membrane protein; OXA1L, mitochondrial inner membrane protein; OXA1-like protein; oxidase (cytochrome c) assembly 1-like; oxidase assembly 1-like; oxidase assembly 1-like protein; zgc:163091; zgc:171949 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 5018 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q15070 | |
| OXA1L | |
| Synthetic peptide directed towards the C-terminal of human OXA1L (aa 371-420). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction