Learn More
Melford Oxacillin Sodium Salt Monohydrate
Description
Oxacillin Sodium Salt Monohydrate is a significant compound playing a critical role in microbiology and pharmaceutical research.
In microbiology, Oxacillin Sodium Salt Monohydrate is extensively employed to investigate bacterial susceptibility and resistance patterns. It is often used in laboratory settings to assess the effectiveness of antibiotics against various bacterial strains. This research aids in understanding the complex dynamics of microbial biology, including mechanisms of resistance, and contributes to the development of novel antibacterial strategies.
Within pharmaceutical research, this compound serves as a reference standard for antibiotic formulation and efficacy testing, supporting the development of new antibiotics and strategies to combat antibiotic-resistant bacteria.
Specifications
Specifications
| Chemical Name or Material | Oxacillin Sodium Salt Monohydrate |
| CAS | 7240-38-2 |
| Quantity | 1 g |
| Molecular Formula | C19H18N3NaO5S H2O |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.