Learn More
Oxalic-13C2 Acid Disodium Salt, TRC
Specifications
Specifications
| Chemical Name or Material | Oxalic-13C2 Acid Disodium Salt |
| CAS | 260429-91-2 |
| Molecular Formula | 13C2Na2O4 |
| Quantity | 100 mg |
| InChI Formula | InChI=1S/C2H2O4.2Na/c3-1(4)2(5)6;;/h(H,3,4)(H,5,6);;/q;2*+1/p-2/i1+1,2+1;; |
| SMILES | O=[13C](O[Na])[13C](O[Na])=O |
| Molecular Weight (g/mol) | 135.98 |
| Formula Weight | 135.9659 |