Learn More
OXCT1 Rabbit, anti-Human, CoraLite Plus 488, Clone: 4K12, Proteintech
Rabbit Recombinant Monoclonal Antibody
Brand:  Proteintech CL488-81011-100UL
 View more versions of this product
Product Code. 18066195
  • € 450.00 / 100µL
Description
The OXCT1 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human OXCT1. This antibody recognizes human, mouse, rat antigen. The OXCT1 antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
OXCT1
4K12
CoraLite Plus 488
Rabbit
Primary
Human, Mouse, Rat
150 kDa
IgG
Immunofluorescence, Flow Cytometry (Intracellular Staining)
1000 μg/mL
3 oxoacid CoA transferase 1, OXCT, OXCT1, SCOT, SCOT s
100 μL
5019
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.