Learn More
OXCT1 Rabbit, anti-Human, CoraLite Plus 488, Clone: 4K12, Proteintech
Rabbit Recombinant Monoclonal Antibody
Brand: Proteintech CL488-81011-100UL
Description
The OXCT1 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human OXCT1. This antibody recognizes human, mouse, rat antigen. The OXCT1 antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite™ plus 488.
Specifications
| OXCT1 | |
| 4K12 | |
| CoraLite Plus 488 | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat | |
| 150 kDa | |
| IgG |
| Immunofluorescence, Flow Cytometry (Intracellular Staining) | |
| 1000 μg/mL | |
| 3 oxoacid CoA transferase 1, OXCT, OXCT1, SCOT, SCOT s | |
| 100 μL | |
| 5019 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |