missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OXCT2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17122-1-AP-20UL
This item is not returnable.
View return policy
Description
Key enzyme for ketone body catabolism. Transfers the CoA moiety from succinate to acetoacetate. Formation of the enzyme-CoA intermediate proceeds via an unstable anhydride species formed between the carboxylate groups of the enzyme and substrate.Specifications
| OXCT2 | |
| Polyclonal | |
| Unconjugated | |
| OXCT2 | |
| 3 oxoacid CoA transferase 2, 3 oxoacid CoA transferase 2A, FKSG25, OXCT2, SCOT T | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 366463, 64059, 64064 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.29 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9BYC2, Q9JJN4 | |
| OXCT2, Oxct2a | |
| OXCT2 Fusion Protein Ag10810 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction