missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OXER1/5-oxo-ETE GPCR Antibody, Novus Biologicals™
Mouse Polyclonal Antibody
Brand: Novus Biologicals H00165140-B01P
This item is not returnable.
View return policy
Description
OXER1/5-oxo-ETE GPCR Polyclonal antibody specifically detects OXER1/5-oxo-ETE GPCR in Human samples. It is validated for Western Blot
Specifications
| OXER1/5-oxo-ETE GPCR | |
| Polyclonal | |
| PBS (pH 7.4) | |
| 5-oxo-ETE acid G-protein-coupled receptor 1,5-oxo-ETE G-protein coupled receptor, GPCR, GPR170G-protein coupled receptor R527, G-protein coupled receptor 170, oxoeicosanoid (OXE) receptor 1, oxoeicosanoid receptor 1, TG1019G-protein coupled receptor TG1019 | |
| OXER1 (NP_683765.1, 1 a.a. - 423 a.a.) full-length human protein. MLCHRGGQLIVPIIPLCPEHSCRGRRLQNLLSGPWPKQPMELHNLSSPSPSLSSSVLPPSFSPSPSSAPSAFTTVGGSSGGPCHPTSSSLVSAFLAPILALEFVLGLVGNSLALFIFCIHTRPWTSNTVFLVSLVAADFLLISNLPLRVDYYLLHETWRFGAAACKVNLFMLSTNRTASVVFLTAIALNRYLKVVQPHHVLSRASVGAAARVAGGLWVGILLLNGHLLLSTFSGPSCLSYRVGTKPSASLRWHQALYLLEFFLPLALILFAIVSIGLTIRNRGLGGQAGPQRAMRVLAMVVAVYTICFLPSIIFGMASMVAFWLSACRSLDLCTQLFHGSLAFTYLNSVLDPVLYCFSSPNFLHQSRALLGLTRGRQGPVSDESSYQPSRQWRYREASRKAEAIGKLKVQGEVSLEKEGSSQG | |
| 0.05 mg | |
| GPCR | |
| 165140 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| NP_683765.1 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction