missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OXLD1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA563362
This item is not returnable.
View return policy
Description
Immunogen sequence: FLQRHHPGAQ APDGRRKFGT DHVEVGSQAG ADGTRPPKAS LPPELQPPTN C Highest antigen sequence identity to the following orthologs: Mouse - 46%, Rat - 49%.
Specifications
| OXLD1 | |
| Polyclonal | |
| Unconjugated | |
| OXLD1 | |
| C17orf90; oxidoreductase like domain containing 1; oxidoreductase-like domain containing 1; oxidoreductase-like domain-containing protein 1; OXLD1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 339229 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5BKU9 | |
| OXLD1 | |
| Recombinant protein corresponding to Human OXLD1. Recombinant protein control fragment (Product #RP-101805). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction