missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OXNAD1 Antibody (OTI1D1), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73175V
This item is not returnable.
View return policy
Description
OXNAD1 Monoclonal specifically detects OXNAD1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
| OXNAD1 | |
| Monoclonal | |
| DyLight 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| OXNAD1 | |
| Full length recombinant protein of human OXNAD1 (NP612390)produced in HEK293 cell. | |
| 0.1 mL | |
| 92106.0 | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| OTI1D1 | |
| Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| MGC15763, oxidoreductase NAD-binding domain containing 1, oxidoreductase NAD-binding domain-containing protein 1 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction