Learn More
Thermo Scientific™ Oxoid™ Anaerobic Low Temperature Catalyst
Click to view available options
Quantity:
1 Ea.
Unit Size:
Pack of 5
Description
Oxoid Anaerobic Low Temperature Catalyst is a safety catalyst designed to stay cooler than traditional catalysts.
- To be changed every 30 cycles
Specifications
Specifications
| Type | Anaerobic Low Temperature Catalyst |
| Quantity | 1 Ea. |