missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Bile Salts

Product Code. p-200112604
Click to view available options
Quantity:
250 g
Unit Size:
250g
This item is not returnable. View return policy

Product Code. 10128872

Brand: Thermo Scientific™ LP0055J

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Oxoid Bile Salts is a standardized ox-bile extract used as a selective inhibitory agent in culture media.

Oxoid Bile Salts is a standardized ox-bile extract used as a selective inhibitory agent in culture media.

Normal working concentration 0.5% w/v. Each vial supplements 500mL of medium.
TRUSTED_SUSTAINABILITY

Specifications

Description Bile Salts
Form Powder
Product Type Bile Salts
Quantity 250 g
compliance-icons
Product Identifier
  • BILE SALTS L55
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Sodium Taurocholate,Sodium Taurodeoxycholate, Sodium cholate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.