Learn More
Thermo Scientific™ Oxoid™ Bile Salts No. 3
Product Code. p-200112588
Click to view available options
Quantity:
250 g
Unit Size:
250g
Product Code. 10423863
Brand: Thermo Scientific™ LP0056J
The Oxoid Bile Salts No. 3 is a specifically refined fraction of bile acid salts.
Oxoid Bile Salts No. 3 is a specifically refined fraction of bile acid salts.
Normal working concentration 0.15% w/v. Each vial supplements 500mL of medium.
TRUSTED_SUSTAINABILITY
Specifications
Description Bile Salts No. 3
Form Powder
Product Type Bile Salts
Quantity 250 g
compliance-icons
Product Identifier
  • BILE SALTS No 3 L56
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : (3.alpha.,5.beta.,7.alpha.,12.alpha.)-3,7,12-trihydroxycholan-24-oic acid