Learn More
Thermo Scientific™ Oxoid™ Burkholderia Cepacia Selective Supplement
Click to view available options
Quantity:
10/Pk.
Unit Size:
Pack of 10
Description
Oxoid Burkholderia Cepacia Selective Supplement is used for the isolation of Burkholderia cepacia.
- Add to Burkholderia Cepacia Agar Base, Part No. CM0995B
- Each vial supplements 500mL of medium
Specifications
Specifications
| Description | Burkholderia Cepacia Selective Supplement |
| Form | Freeze-dried |
| Product Type | Selective Supplement |
| Quantity | 10/Pk. |
| Yield | Each vial for 500mL medium |
| Species | Burkholderia cepacia |