missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Gamma-Irradiated Tryptone Soya Broth (Dehydrated)

Product Code. 10198002
Click to view available options
Quantity:
500 g
5 kg
Unit Size:
500g
5kg
This item is not returnable. View return policy

Product Code. 10198002

Brand: Thermo Scientific™ CM1016B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cultivate a wide range of organisms with this gamma-irradiated medium that is particularly suitable for the Media Fill Tests.

Cultivate a wide range of organisms with nutritious Thermo Scientific™ Oxoid™ Gamma-Irradiated Tryptone Soya Broth (TSB) (Dehydrated) which has been given a sterilizing dose of irradiation. Gamma-Irradiated Tryptone Soya Broth is particularly suitable for the microbiological validation of aseptic filling (Media Fill Test).

Media Fill Test is a validation method used to assess the performance of aseptic processing in the pharmaceutical industry. Pharmaceutical products are substituted by Gamma irradiated TSB and the medium is subjected to exactly the same conditions as the products, including filling and closing, to ensure that there is no microbial contamination occurring during the process.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand

TRUSTED_SUSTAINABILITY

Specifications

Description Tryptone Soya Broth, Gamma-Irradiated
Form Powder
Product Type Dehydrated Culture Media
Quantity 500 g

For Laboratory Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.