Learn More
Thermo Scientific™ Oxoid™ Half Fraser Supplement
Description
Oxoid Half Fraser Supplement is used for the enrichment of Listeria species.
- Add to Fraser Broth Base, Part No. CM0895B
Specifications
Specifications
| Description | Half Fraser Supplement |
| Form | Freeze-dried |
| Product Type | Microbiology Media |
| Quantity | 10/Pk. |
| Yield | Each vial for 225mL medium |
| Species | Listeria |