Learn More
Thermo Scientific™ Oxoid™ Peptone Bacteriological Neutralized
Click to view available options
Quantity:
500 g
Unit Size:
500g
Description
Oxoid Peptone Bacteriological Neutralized is the neutralized form of Peptone Bacteriological.
Specifications
Specifications
| Description | Peptone Bacteriological Neutralized |
| Form | Powder |
| Product Type | Peptone |
| Quantity | 500 g |