Learn More
Thermo Scientific™ Oxoid™ Prepared MacConkey Broth Bottled Media
Product Code. 12593936
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12593936 100 mL Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12593936 Supplier Thermo Scientific™ Supplier No. BO1124M
Oxoid™ Prepared MacConkey Broth Bottled Media, 100 mL, containing neutral red for the detection of coliform organisms in water and milk examination.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Microbiology Media
Quantity 100 mL