Learn More
Thermo Scientific™ Oxoid™ Sabouraud Dextrose Agar + Chloramphenicol Slope
Click to view available options
Fill Weight:
15-15.3 g
Format:
Universal bottle
Unit Size:
Pack of 24
Description
Sabouraud Dextrose Agar and Chloramphenicol Slope (BO0422F) is an acidic pH medium for the detection of fungi and yeasts from clinical samples.
Specifications
Specifications
| Color | Straw |
| Content And Storage | 2 to 25°C |
| Format | Universal bottle |
| Manufacturing Origin | United Kingdom |
| Product Type | Agar |
| Fill Weight | 15-15.3 g |
| Packaging Type | Twenty-four universal bottles with screw cap closures in a box. |
| Quantity | 24/Pk. |