missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OXPAT Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB110-60511FR
This item is not returnable.
View return policy
Description
OXPAT Polyclonal antibody specifically detects OXPAT in Human, Mouse samples. It is validated for Western Blot, Simple Western
Specifications
| OXPAT | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Simple Western | |
| DyLight 680 | |
| lipid storage droplet protein 5, LSDA5, LSDP5, MLDP, OXPAT, perilipin 5, perilipin-5 | |
| A synthetic peptide made to a C-terminal region of the mouse OXPAT protein, within residues 400-463. [UniProt# Q8BVZ1] | |
| 0.1 mL | |
| Diabetes Research, Lipid and Metabolism, Lipid Droplets | |
| 440503 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction