missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OXR1 Antibody, mFluor Violet 450 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97444MFV450
This item is not returnable.
View return policy
Description
OXR1 Polyclonal antibody specifically detects OXR1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| OXR1 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin | |
| FLJ10125, FLJ38829, FLJ40849, FLJ41673, FLJ42450, FLJ45656, oxidation resistance 1, oxidation resistance protein 1, putative protein product of Nbla00307 | |
| Produced in rabbits immunized with E. coli-derived Human OXR1 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 55074 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction