missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OXR1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 292.00 - € 572.00
Specifications
| Antigen | OXR1 |
|---|---|
| Dilution | Western Blot 0.04-0.4 ug/mL, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18472731
|
Novus Biologicals
NBP1-86391 |
0.1 mL |
€ 572.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18425731
|
Novus Biologicals
NBP1-86391-25ul |
25 μL |
€ 292.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
OXR1 Polyclonal antibody specifically detects OXR1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| OXR1 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| FLJ10125, FLJ38829, FLJ40849, FLJ41673, FLJ42450, FLJ45656, oxidation resistance 1, oxidation resistance protein 1, putative protein product of Nbla00307 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: EEYGIMCPMEEVMSAAMYKEILDSKIKESLPIDIDQLSGRDFCHSKKMTGSNTEEIDSRIRDAGNDSASTAPRSTEESLSEDVFTESELSPIR | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot 0.04-0.4 ug/mL, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol | |
| 55074 | |
| IgG | |
| Immunogen affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title