missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OXSR1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-77321
This item is not returnable.
View return policy
Description
OXSR1 Polyclonal specifically detects OXSR1 in Human samples. It is validated for Immunohistochemistry, Immunoprecipitation, Western Blot (Negative).
Specifications
| OXSR1 | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| EC 2.7.11, EC 2.7.11.1, KIAA1101OSR1serine/threonine-protein kinase OSR1, Oxidative stress-responsive 1 protein, oxidative-stress responsive 1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
| Immunohistochemistry, Immunoprecipitation, Western Blot | |
| 1.0 mg/mL | |
| Immunohistochemistry, Immunoprecipitation 1:10-1:500, Western Blot (Negative) | |
| NP_005100.1 | |
| OXSR1 | |
| The immunogen recognized by this antibody maps to a region between residue 325 and 375 of human oxidative-stress responsive 1 using the numbering given in entry NP_005100.1 (GeneID 9943). | |
| 100 μL | |
| Protein Kinase | |
| 9943 | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction