missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OXSR1 Antibody (OTI1F3), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73177AF647
This item is not returnable.
View return policy
Description
OXSR1 Monoclonal specifically detects OXSR1 in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| OXSR1 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| OXSR1 | |
| Full-length protein expressed in 293T cell transfected with human OXSR1 expression vector. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Canine, Primate, Monkey | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI1F3 | |
| Western Blot, Immunohistochemistry | |
| EC 2.7.11, EC 2.7.11.1, KIAA1101OSR1serine/threonine-protein kinase OSR1, Oxidative stress-responsive 1 protein, oxidative-stress responsive 1 | |
| Mouse | |
| Affinity Purified | |
| Protein Kinase | |
| 9943.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction