Learn More
OXT Monoclonal Antibody (OTI5G4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA807052
Encompass_Preferred
 View more versions of this product
Product Code. 17517413
  • € 540.87 / 100µL
Description
There are two proteins encoded by this gene, oxytocin and neurophysin I. Oxytocin is posterior pituitary hormone which is synthesized as an inactive precursor in the hypothalamus along with its carrier protein neurophysin I. Together with neurophysin, it is packaged into neurosecretory vesicles and transported axonally to the nerve endings in the neurohypophysis, where it is either stored or secreted into the bloodstream. The precursor seems to be activated while it is being transported along the axon to the posterior pituitary. This hormone contracts smooth muscle during parturition and lactation. It is also involved in cognition, tolerance, adaptation and complex sexual and maternal behaviour, as well as in the regulation of water excretion and cardiovascular functions.
Specifications
Specifications
OXT
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P01178
OXT
Human recombit protein fragment corresponding to amino acids 20-125 of human OXT produced in E.coli.
100 μL
Primary
Human
Antibody
IgG2b
Western Blot
OTI5G4
Unconjugated
OXT
OT, OT-NPI, OXT-NPI
Mouse
Affinity Chromatography
RUO
5020
-20° C, Avoid Freeze/Thaw Cycles