Learn More
Lenz Laborglasintrumente Oxygen Determination Bottles Acc. to Winkler
Product Code. 10518431
Change view
Click to view available options
Standard Taper Joint:
NS 14/23
NS 19/26
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Taper Joint unitSize
10518431 NS 19/26 Each
10578421 NS 14/23 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10518431 Supplier Lenz Laborglasintrumente Supplier No. 05605049
Only null left
Add to basket
Add to basket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • For the determination of the solved oxygen in water; capacity engraving at the bottle (accuracy: ± 0,01 ml)
  • With inscription label
  • With solid, angled grinded NS glass stopper (can be fixed by clamp)
  • Each bottle is individually adjusted with its stopper and both are marked with the same number
  • Material: soda lime glass
Specifications
Material Soda lime glass
Capacity (Metric) 250 to 300 mL
Standard Taper Joint NS 19/26