missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oxyntomodulin Antibody (46), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61947IR
This item is not returnable.
View return policy
Description
Oxyntomodulin Monoclonal antibody specifically detects Oxyntomodulin in Human samples. It is validated for ELISA
Specifications
| Oxyntomodulin | |
| Monoclonal | |
| DyLight 755 | |
| GCG, GLP1, GLP2, GRPP | |
| Synthetic oxyntomodulin 31-37 coupled to carrier. | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| ELISA | |
| 46 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction