missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oxyquinoline Sulfate, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Oxyquinoline Sulfate
Specifications
Specifications
| Chemical Name or Material | 8-Hydroxyquinoline sulfate |
| CAS | 134-31-6 |
| Analyte or Component Names | 8-Hydroxyquinoline sulfate |
| Shipping Condition | Room Temperature |
| Molecular Formula | 2 C9 H7 N O . H2 O4 S |
| Certifications/Compliance | ISO 17034 |
| Synonym | 8-Quinolinol Sulfate (2:1), 8-Quinolinol, sulfate (2:1) (salt) (8CI,9CI), 8-Hydroxyquinoline sulfate, 8-Hydroxyquinoline sulfuric acid salt, 8-Hydroxyquinoline sulphate, 8-Quinolinol sulfate, 8-Quinolinol sulfate (salt), 8-Quinolinol sulphate, Chinosol, Criptonol, Cryptonol, Hydroxyquinoline sulfate, Khinozol, Octofen, Oxine sulfate, Oxyquinol, Oxyquinoline sulfate, Quinolin-8-ol sulfate (2:1), Quinosol, Quinosol extra, Solvochin-Extra, Sunoxol, Superol |
| InChI Formula | InChI=1S/2C9H7NO.H2O4S/c2*11-8-5-1-3-7-4-2-6-10-9(7)8;1-5(2,3)4/h2*1-6,11H;(H2,1,2,3,4) |
| SMILES | Oc1cccc2cccnc12.Oc3cccc4cccnc34.OS(=O)(=O)O |
| IUPAC Name | quinolin-8-ol;sulfuric acid |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction