Learn More
p-Anisidine hydrochloride, 99%
Product Code. p-61033 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
Product Code. 10202582
Brand: Thermo Scientific Acros 401300050
Request Bulk or Custom Format
CAS: 20265-97-8 | C7H9NO·HCl | 159.61 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 20265-97-8
Molecular Formula C7H9NO·HCl
Molecular Weight (g/mol) 159.61
MDL Number MFCD00036388
InChI Key VQYJLACQFYZHCO-UHFFFAOYSA-N
Synonym p-anisidine hydrochloride, 4-methoxyaniline hydrochloride, unii-7c4r75n8vg, benzenamine, 4-methoxy-, hydrochloride, p-anisidine.hcl, p-anisidine hcl, p-anisidinium chloride, 4-anisidine hydrochloride, p-anisylamine hydrochloride, p-aminoanisole hydrochloride
PubChem CID 2734956
IUPAC Name 4-methoxyaniline;hydrochloride
SMILES COC1=CC=C(C=C1)N.Cl
Specifications
Melting Point 216.0°C to 219.0°C
Color Brown-Gray to White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula NH2C6H4OCH3·HCl
Quantity 5 g
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol
Formula Weight 159.61
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material p-Anisidine hydrochloride, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • p-Anisidine hydrochloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P361-Take off immediately all contaminated clothing.