missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P-Cadherin Antibody (106020), PE/Cy5.5, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB761PECY55
This item is not returnable.
View return policy
Description
P-Cadherin Monoclonal antibody specifically detects P-Cadherin in Human, Mouse samples. It is validated for Flow Cytometry
Specifications
| P-Cadherin | |
| Monoclonal | |
| PE-Cyanine5.5 | |
| PBS | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Flow Cytometry | |
| 106020 | |
| Flow Cytometry | |
| cadherin 3, P-cadherin (placental), cadherin 3, type 1, P-cadherin (placental), cadherin-3, CDHPcalcium-dependent adhesion protein, placental, HJMD, PCADP-cadherin, Placental cadherin | |
| Mouse myeloma cell line NS0-derived recombinant mouse P-Cadherin, Glu100-Gly647, Accession # Q8BSL6 | |
| 0.1 mL | |
| Cancer | |
| 1001 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction