missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P-Cadherin Antibody (12H6), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-54535C
This item is not returnable.
View return policy
Description
P-Cadherin Monoclonal antibody specifically detects P-Cadherin in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, CyTOF
Specifications
| P-Cadherin | |
| Monoclonal | |
| DyLight 650 | |
| cadherin 3, P-cadherin (placental), cadherin 3, type 1, P-cadherin (placental), cadherin-3, CDHPcalcium-dependent adhesion protein, placental, HJMD, PCADP-cadherin, Placental cadherin | |
| Recombinant human P-Cadherin protein (Uniprot: P22223) | |
| 100 μg | |
| Cancer | |
| 1001 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, Immunofluorescence, CyTOF | |
| 12H6 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction