missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P-Cadherin Antibody (M596), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-23436
This item is not returnable.
View return policy
Description
P-Cadherin Monoclonal antibody specifically detects P-Cadherin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| P-Cadherin | |
| Monoclonal | |
| Unconjugated | |
| PBS, 1 mg/mL BSA and 50% Glycerol | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunofluorescence | |
| M596 | |
| Western Blot 1:1000, Immunocytochemistry/ Immunofluorescence 1:100 | |
| cadherin 3, P-cadherin (placental), cadherin 3, type 1, P-cadherin (placental), cadherin-3, CDHPcalcium-dependent adhesion protein, placental, HJMD, PCADP-cadherin, Placental cadherin | |
| Clone (M596) was generated from a recombinant protein containing amino acids in the N-terminal region of human P-Cadherin. This sequence is highly conserved in mouse and rat P-cadherin. [UniProt# P22223] | |
| 0.1 mL | |
| Cancer | |
| 1001 | |
| Store at -20°C. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction