Learn More
Invitrogen™ P-cadherin Recombinant Rabbit Monoclonal Antibody (8J5J8)
Rabbit Recombinant Monoclonal Antibody
€ 176.00 - € 436.00
Specifications
| Antigen | P-cadherin |
|---|---|
| Clone | 8J5J8 |
| Concentration | 0.86 mg/mL |
| Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
| Applications | ELISA, Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30284110
|
Invitrogen™
MA558201 |
20 μL |
€ 176.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30283892
|
Invitrogen™
MA558202 |
100 μL |
€ 436.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Immunogen sequence: DWVVAPISVP ENGKGPFPQR LNQLKSNKDR DTKIFYSITG PGADSPPEGV FAVEKETGWL LLNKPLDREE IAKYELFGHA VSENGASVED PMNISIIVTD QNDHKPKFTQ DTFRGSVLEG VLPGTSVMQV TATDEDDAIY TYNGVVAYSI HSQEPKDPHD LMFTIHRSTG TISVISSGLD REKVPEYTLT IQATDMDGDG STTTAVAVVE ILDANDNAPM FDPQKYEAHV PENAVGHEVQ RLTVTDLDAP NSPAWRATYL IMGGDDGDHF TITTHPESNQ GILTTRKGLD FEAKNQHTLY VEVTNEAPFV LKLPTSTATI VVHVEDVNEA PVFVPPSKVV EVQEGIPTGE PVCVYTAEDP DKENQKISYR ILRDPAGWLA MDPDSGQVTA VGTLDREDEQ FVRNNIYEVM VLAMDNGSPP TTGTGTLLLT LIDVNDHGPV PEPRQITICN QSPVRQVLNI TDKDLSPHTS PFQAQLTDDS DIYWTAEVNE EGDTVVLSLK KFLKQDTYDV HLSLSDHGNK EQLTVIRATV CDCHGHVETC PGPWKGG.
P-cadherin, also known as Cadherin-3, is a Type 1 cadherin protein that belongs to the cadherin superfamily. Type 1 cadherins are single-pass transmembrane proteins that have 5 extracellular cadherin repeats and an intracellular domain that binds p120-catenin and beta-catenin. Cadherins are calcium-dependent cell-cell adhesion glycoproteins responsible for a range of processes including development, wound healing, cell-cell signaling, cell growth and differentiation. P-cadherin is expressed in human fetal structures, and in adult tissues such as the basal layer of the epidermis, breast, prostate, ovary, cervix, hair follicle, and corneal endothelium. P-cadherin expression has also been reported on embryonic stem cells, and stem cells of the normal mammary gland, and hair follicle. Overexpression of P-cadherin has been associated with poor prognosis in breast, prostate, ovary, colon, and stomach carcinomas. Mutations in CDH3 are associated with congential hypotrichosis with juvenile macular dystrophy.Specifications
| P-cadherin | |
| 0.86 mg/mL | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| P22223 | |
| 1001 | |
| Recombinant protein (or fragment). | |
| Antibody |
| 8J5J8 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human | |
| Cdh3 | |
| AI385538; CADH3; cadherin 15, M-cadherin (myotubule); cadherin 3; cadherin 3, type 1, P-cadherin (placental); cadherin-3; Cadp; calcium-dependent adhesion protein, placental; CDH3; CDHP; HJMD; PCAD; P-cadherin; Placental cadherin; Placental-cadherin; RPE-specific cadherin | |
| Cdh3 | |
| Primary | |
| Protein A |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.