Learn More
P-glycoprotein Mouse anti-Human, Brilliant Violet 711, Clone: 17F9, BD Optibuild™
Mouse Monoclonal Antibody
Brand: BD Biosciences 742851
Description
The 17F9 monoclonal antibody specifically binds to the 170-180 kDa transmembrane glycoprotein (P-glycoprotein), a product of the multidrug resistance-1 (MDR1) gene. This glycoprotein is expressed on MDR positive cells and has been reported to be expressed on many normal tissues, such as adrenal glands and endothelium, in the brain and skin. P-glycoprotein is known to impart drug resistance to cells by pumping many anti-cancer drugs out of the cytoplasm. 17F9 antibody is able to partially block the binding of UIC2 antibody (another MDR-specific monoclonal antibody).
The antibody was conjugated to BD Horizon™ BV711 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 711nm. BD Horizon BV711 can be excited by the violet laser and detected in a filter used to detect Cy™5.5 / Alexa Fluor™ 700-like dyes (eg, 712/20nm filter). Due to the excitation and emission characteristics of the acceptor dye, there may be moderate spillover into the Alexa Fluor™ 700 and PerCP-Cy5.5 detectors. However, the spillover can be corrected through compensation as with any other dye combination. BV711 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 711 nm. This dye offers a very bright choice for the violet laser. Due to the excitation and emission characteristics of the acceptor dye, there may be moderate spillover into the Alexa Fluor™ 700 and PerCP-Cy5.5 detectors. BV711 will also have moderate spillover into the BD Horizon Brilliant™ Violet 786 detector.
Specifications
| P-glycoprotein | |
| 17F9 | |
| Brilliant Violet 711 | |
| CD243, ABCB1, ABC20, CLCS, GP170, MDR1, P-GP, PGY1 | |
| Human MDR1 Transfected Cell Line | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Monoclonal | |
| 0.2mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG2b κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.