Learn More
Bohlender™ P-MRK Clamp Nut
Suitable for use with P-MRK Magnetic Stirrer Heads.
Brand: Bohlender™ C925-01
This item is not returnable.
View return policy
Description
- Manufactured from a PTFE-PEEK compound
- For use with P-MRK stirrer heads with ground joint NS 29/32 for shaft O.D. 8 mm as well as P-MRK stirrer heads with ground joint NS 29/32 or NS 45/40 for shaft O.D. 10 mm and stirrer heads with Flat Flange DN 40 or DN 50 for shaft O.D. 10 mm
- FDA conformity
- Provided in packs of 3
Specifications
| Nut | |
| PTFE-PEEK Compound | |
| Magnetic Stirrer Heads | |
| White |