missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bohlender™ P-MRK O-Rings
Click to view available options
Quantity:
3/pk
Unit Size:
Pack of 3
Description
- Manufactured from PFA/silicone
- For use with P-MRK stirrer heads of various sizes
- FDA conformity
- Provided in packs of 3
Specifications
Specifications
| For Use With (Equipment) | Stirrer heads with ground joint NS 29/32 or NS 45/40 for shaft O.D. 10 mm and heads with flat flange DN 40 or DN 50 for shaft O.D. 10 mm |
| Quantity | 3/pk |
| Product Type | O-Ring |
| Material | PFA/silicone |
| Color | Red |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction