Learn More
Melford P-Nitrophenyl-a-D-galactopyranoside
Product Code. 30018631
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018631 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30018631 Supplier Melford Supplier No. N820601.0
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.
Free PNP: <500 ppm
Specifications
Quantity 1 g