missing translation for 'onlineSavingsMsg'
Learn More

p-Nitrophenyl Beta-D-Laminaribioside, TRC

Product Code. 30475751
Change view
Click to view available options
Quantity:
2 mg
Unit Size:
2mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30475751 2 mg 2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30475751 Supplier Toronto Research Chemicals Supplier No. TRCN5038752MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material p-Nitrophenyl beta-D-Laminaribioside
CAS 26255-70-9
Molecular Formula C18 H25 N O13
Quantity 2 mg
Synonym β-D-Glucopyranoside','4-nitrophenyl 3-O-β-D-glucopyranosyl-, Glucopyranoside','p-nitrophenyl 3-O-β-D-glucopyranosyl-','β-D- (8CI), 4-Nitrophenyl 3-O-β-D-glucopyranosyl-β-D-glucopyranoside, 4-Nitrophenyl-β-laminaribioside, p-Nitrophenyl β-D-laminaribioside
InChI Formula InChI=1S/C18H25NO13/c20-5-9-11(22)13(24)14(25)17(30-9)32-16-12(23)10(6-21)31-18(15(16)26)29-8-3-1-7(2-4-8)19(27)28/h1-4,9-18,20-26H,5-6H2/t9-,10-,11-,12-,13+,14-,15-,16+,17+,18?/m1/s1
SMILES OC[C@H]1O[C@@H](O[C@H]2[C@H](O)[C@@H](CO)OC(Oc3ccc(cc3)[N+](=O)[O-])[C@@H]2O)[C@H](O)[C@@H](O)[C@@H]1O
IUPAC Name (2S,3R,4S,5S,6R)-2-[(2R,3R,4S,5R)-3,5-dihydroxy-2-(hydroxymethyl)-6-(4-nitrophenoxy)oxan-4-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular Weight (g/mol) 463.39
Formula Weight 463.1326
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.