missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ P-Nitrophenyl-α-D-Galactopyranoside, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
500mg
Unit Size:
500mg
Specifications
Specifications
| Form | Powder |
| Quantity | 500mg |
| Format | White Powder |
| Molecular Weight (g/mol) | 301.3 |
| Storage Requirements | 0°C, protect from light |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction