missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P-Rex1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98883-50ul
This item is not returnable.
View return policy
Description
P-Rex1 Polyclonal antibody specifically detects P-Rex1 in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| P-Rex1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunoprecipitation 1-4 μL/mg of lysate | |
| KIAA1415, phosphatidylinositol 34,5-trisphosphate-dependent Rac exchanger 1 protein, phosphatidylinositol-34,5-trisphosphate-dependent Rac exchange factor 1, ptdIns(34,5)-dependent Rac exchanger 1 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the human P-Rex1. | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 57580 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction