missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P-Selectin/CD62P Antibody (001), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-90703CL1
This item is not returnable.
View return policy
Description
P-Selectin/CD62P Monoclonal antibody specifically detects P-Selectin/CD62P in Mouse samples. It is validated for ELISA
Specifications
| P-Selectin/CD62P | |
| Monoclonal | |
| CoraFluor 1 | |
| CD62P antigen, FLJ45155, PADGEMantigen CD62), selectin P (granule membrane protein 140kDa, antigen CD62) | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse P-Selectin/CD62P (Uniprot#: Q01102; Met1-Ala709). | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
| ELISA | |
| 001 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 6403 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction