missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P-Selectin/CD62P Antibody (Psel.KO.2.12), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-65391CL1
This item is not returnable.
View return policy
Description
P-Selectin/CD62P Monoclonal antibody specifically detects P-Selectin/CD62P in Goat,Human,Rat,Sheep samples. It is validated for Flow Cytometry,Immunohistochemistry,Immunoprecipitation,Immunohistochemistry (Frozen)
Specifications
| P-Selectin/CD62P | |
| Monoclonal | |
| CoraFluor 1 | |
| CD62P antigen, FLJ45155, PADGEMantigen CD62), selectin P (granule membrane protein 140kDa, antigen CD62) | |
| CD62P transfected 300.19 cells | |
| 0.1 mL | |
| Primary | |
| Goat, Human, Rat, Sheep | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen) | |
| Psel.KO.2.12 | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 6403 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction