missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P-Selectin/CD62P Antibody (Psel.KO.2.7), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-65392AF647
This item is not returnable.
View return policy
Description
P-Selectin/CD62P Monoclonal specifically detects P-Selectin/CD62P in Human, Mouse, Rat, Bovine, Equine, Feline, Goat, Sheep samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
| P-Selectin/CD62P | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CD62P antigen, FLJ45155, PADGEMantigen CD62), selectin P (granule membrane protein 140kDa, antigen CD62) | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 6403 | |
| Human, Mouse, Rat, Bovine, Equine, Feline, Goat, Sheep | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| Psel.KO.2.7 | |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen | |
| P16109 | |
| SELP | |
| P-selectin transfected 300.19 cells. | |
| 0.1 mL | |
| Primary | |
| NB100-65392 recognizes the P-Selectin cell surface antigen, a 140kD glycoprotein. P-Selectin is expressed by activated platelets and endothelial cells, and plays an important role in adhesive processes between leucocytes and endothelial cells. | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction