missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P-Selectin/CD62P Antibody (Psel.KO.2.7), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-65392PCP
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
P-Selectin/CD62P Monoclonal specifically detects P-Selectin/CD62P in Human, Mouse, Rat, Bovine, Equine, Feline, Goat, Sheep samples. It is validated for Flow Cytometry.
Specifikationer
| P-Selectin/CD62P | |
| Monoclonal | |
| PerCP | |
| P16109 | |
| SELP | |
| P-selectin transfected 300.19 cells. | |
| 0.1 mL | |
| Primary | |
| NB100-65392 recognizes the P-Selectin cell surface antigen, a 140kD glycoprotein. P-Selectin is expressed by activated platelets and endothelial cells, and plays an important role in adhesive processes between leucocytes and endothelial cells. | |
| Store at 4C in the dark. | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen) | |
| Psel.KO.2.7 | |
| Flow Cytometry | |
| CD62P antigen, FLJ45155, PADGEMantigen CD62), selectin P (granule membrane protein 140kDa, antigen CD62) | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 6403 | |
| Human, Mouse, Rat, Bovine, Equine, Feline, Goat, Sheep | |
| Purified |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering