missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P-Selectin/CD62P Rabbit anti-Human, Clone: 1, Novus Biologicals™
Rabbit Monoclonal Antibody
€ 379.00 - € 552.00
Specifications
| Antigen | P-Selectin/CD62P |
|---|---|
| Clone | 1 |
| Dilution | ELISA 1:5000-1:10000 |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18233708
|
Novus Biologicals
NBP2-90156-50UL |
50 μg |
€ 379.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18237276
|
Novus Biologicals
NBP2-90156-100UL |
100 μg |
€ 552.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
P-Selectin/CD62P Monoclonal specifically detects P-Selectin/CD62P in Human samples. It is validated for ELISA.Specifications
| P-Selectin/CD62P | |
| ELISA 1:5000-1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| 6403 | |
| IgG | |
| Protein A purified |
| 1 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Human | |
| CD62P antigen, FLJ45155, PADGEMantigen CD62), selectin P (granule membrane protein 140kDa, antigen CD62) | |
| Recombinant Human P-Selectin/CD62P protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title