missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ P-Selectin Polyclonal Antibody

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA588593

Product Code. 16546791

  • € 528.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: FSFNSQCSFH CTDGYQVNGP SKLECLASGI WTNKPPQCLA AQCPPLKIPE RGNMTCLHSA KAFQHQSSCS FSCEEGFALV GPEVVQCTAS GVWTAPAPVC; Positive Samples: Mouse kidney; Cellular Location: Membrane, Single-pass type I membrane protein.

P-selectin (Selectin P, GMP-140, LECAM3, CD62 antigen-like family member) is a 140 kDa type 1 transmembrane glycoprotein, and is one of the most commonly studied proteins that regulate cell rolling. P-selectin is stored in the Weibel-Palade bodies of endothelial cells, as well as in a-granules of platelets. From there, it can be rapidly brought to the cell surface after exposure to thrombin, histamine, complement 5a, Ca21 ionophores, or adenosine diphosphate. P-selectin protein redistributes to the plasma membrane during platelet activation and degranulation and mediates the interaction of activated endothelial cells or platelets with leukocytes. P-selectin plays an important role in adhesive processes between leucocytes and endothelial cells, and is a calcium dependent receptor that binds to sialylated forms of Lewis blood group carbohydrate antigens found on neutrophils and monocytes. P-selectin is constitutively expressed in inflammation and contributes to atherogenesis, thrombosis and tissue destruction. Clinically, P-selectin is used to distinguish heparin induced thrombocytopenia with and without thrombosis.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

P-Selectin
Polyclonal
Unconjugated
SELP
CD62; CD62 antigen-like family member P; CD62P; CD62P antigen; CD62P; CD62 homolog; FLJ45155; GMP140; GMP-140; GMRP; Granule membrane protein 140; granule membrane protein 140kDa; granulocyte membrane protein; Grmp; LECAM3; leukocyte-endothelial cell adhesion molecule 3; PADGEM; Platelet activation dependent granule-external membrane protein; platelet alpha-granule membrane protein; PSEL; PSELECT; P-selectin; P-selectin precursor; RP1-86F14.2; selectin P; selectin P (granule membrane protein 140kDa, antigen CD62); selectin, platelet; Selp
Rabbit
Affinity Chromatography
RUO
20344, 6403
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot
0.69 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P16109, Q01102
SELP
Recombinant fusion protein containing a sequence corresponding to amino acids 220-319 of human CD62P/P-selectin (NP_0029962).
100 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.