missing translation for 'onlineSavingsMsg'
Learn More

Tunicamycin, 95%, Thermo Scientific™

 

Brand:  Thermo Scientific Alfa Aesar J62217.MA

Quantity 10mg

Product Code. 15464629

  • ONLINE EXCLUSIVE PRICE
    € 250.00
    € 225.65 / 10mg
    Save € 24.35
    9% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Tunicamycin has been widely used in the study of glycoprotein synthesis in various biological systems. During protein glycosylation, tunicamycin is noted to be an inhibitor of the transfer of saccharide moieties to dolichol during d
TRUSTED_SUSTAINABILITY
Specifications

Specifications

11089-65-9
10mg
C39H64N4O16
UN3462
14,9819
844.95
Tunicamycin
White to Yellow
Light and moisture sensitive
MFCD00065709
6888090
Soluble in methanol, ethanol, DMSO, or DMF
95%
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.