missing translation for 'onlineSavingsMsg'
Learn More

p120-catenin Antibody (DBM15.55), Novus Biologicals™

Product Code. 30532830 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
0.5 mL
1 mL
Unit Size:
0.1mL
0.5mL
1mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30532830 1 mL 1mL
30532828 0.1 mL 0.1mL
30532829 0.5 mL 0.5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30532830 Supplier Novus Biologicals Supplier No. NBP3451141ml

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

p120-catenin Monoclonal antibody specifically detects p120-catenin in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen p120-catenin
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone DBM15.55
Conjugate Unconjugated
Formulation PBS (pH 7.4), Tween-20, 0.2% BSA
Gene Alias Cadherin-associated Src substrate, CAS, catenin (cadherin-associated protein), delta 1, catenin delta-1, CTNND, KIAA0384P120CAS, p120, p120 catenin, p120(cas), p120(CTN), p120cas, P120CTN
Host Species Mouse
Immunogen Recombinant human p120 Catenin protein fragment
Purification Method Protein A purified
Quantity 1 mL
Regulatory Status RUO
Research Discipline Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1500
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.